Learn More
Thermo Scientific™ PepSwift™ Monolithic Capillary HPLC Columns
Shop All Thermo Scientific ProductsDescription
For fast, high-resolution LC/MS separations in bottom-up protein identification, biomarker discovery, and systems biology applications, use Thermo Scientific™ PepSwift™ Monolithic Capillary LC Columns. Based on a polystyrene divinylbenzene copolymer, the monolithic structure offers a high-quality alternative to traditional microparticulate sorbents, providing important advantages to the chromatographic separation. High-sensitivity proteomics and biotech applications are easily performed using these columns.
Benefit from:
- High-speed peptide and protein separations (<15 min)
- High sensitivity for LC/MS
- High column-to-column reproducibility
- Wide range of column I.D. and lengths available
- Superior lifetime
- Easy column installation using Thermo Scientific™ Dionex™ nanoViper™ Fingertight Fittings
- Precolumns available to preconcentrate and desalt samples
You can also use Thermo Scientific™ PepSwift™ Trap Columns for preconcentration and desalting of samples consisting of peptides and proteins without negatively impacting the chromatographic performance or recovery of the compounds. Change the selectivity of the separation or improve the trapping efficiency by employing various ion-pairing agents in the loading solvent and/or mobile phases. The trap columns are used with Thermo Scientific™ PepSwift™ and ProSwift™ Monolithic Columns, which are for fast, high-resolution LC/MS separations in protein identification, biomarker discovery, and systems biology applications.
Specifications
Specifications
| Column Format | Analytical Column, NanoViper |
| Diameter (Metric) | 0.2 mm |
| Flow Rate | 2 to 3 μL/min |
| Length (Metric) | 50 mm |
| Max. Pressure | 4350 psi (300 bar) |
| Particle Size | Monolith |
| pH | 1 to 10 |
| Pore Size | Monolith |
| Temperature | 80°C |
| Stationary Phase | PSDVB (Polystyrene Divinylbenzene) |
| Show More |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.