missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pepstatin Ammonium, MedChemExpress
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
Description
Pepstatin Ammonium is a specific aspartic protease inhibitor produced by actinomycetes, with IC50s of 4.5 nM, 6.2 nM, 150 nM, 290 nM, 520 nM and 260 nM for hemoglobin-pepsin, hemoglobin-proctase, casein-pepsin, casein-proctase, casein-acid protease and hemoglobin-acid protease, respectively. Pepstatin Ammonium also inhibits HIV protease.
Specifications
Specifications
| Chemical Name or Material | Pepstatin Ammonium |
| Physical Form | Solid |
| Color | White |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C34H66N6O9 |
| For Use With (Application) | COVID-19-anti-virus |
| Synonym | Pepstatin A Ammonium |
| Solubility Information | DMSO : 41.67 mg/mL (59.28 mM; Need ultrasonic) |
| SMILES | CC(C[C@H](NC([C@@H](NC([C@@H](NC(CC(C)C)=O)C(C)C)=O)C(C)C)=O)[C@@H](O)CC(N[C@@H](C)C(N[C@@H](CC(C)C)[C@@H](O)CC(O)=O)=O)=O)C.N |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction