missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Pepstatin A

Product Code. 15865328 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15865328 25 mg 25mg
15855328 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15865328 Supplier Thermo Scientific Chemicals Supplier No. J20037.MB

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Protease inhibitor, inhibits Pepsin, Renin and Cathepsin D.

Applications: Pepstatin A, a protease inhibitor, inhibits Pepsin, Renin and Cathepsin D. Effective concentrations are 10-6 to 10-9M. It does not inhibit Trypsin, Chymotrypsin, Papain or Plasmin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Protease inhibitor, inhibits Pepsin, Renin and Cathepsin D
  • Effective concentrations are 10-6 to 10-9M
  • Does not inhibit Trypsin, Chymotrypsin, Papain or Plasmin
  • Protease inhibitor, inhibits Pepsin, Renin and Cathepsin D
  • MDL: MFCD00060740
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26305-03-3
Molecular Formula C34H63N5O9

Specifications

Concentration 10-6 to 10-9M
Synonym Isovaleryl-val-val-4-amino-3-hydroxy-6-methylheptanoyl-ala-4-amino-3-hydroxy-6-methylheptanoic acid SDS Certificate of Analysis Product Specification Technical Inquiry
Solubility Dissolves in methanol
Form Powder
Quantity 25 mg

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.