missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pepstatin A, Fisher BioReagents
Description
Pepstatin A is an inhibitor for pepsin, renin, cathepsin D, and other acid proteases.
Specifications
Specifications
| Melting Point | 235°C |
| Color | White |
| Melting Range | 230° to 240°C |
| Infrared Spectrum | Conforms to reference |
| Packaging | Amber Glass |
| Quantity | 250 mg |
| Merck Index | 15, 7260 |
| Formula Weight | 685.9 |
| Physical Form | Powder or Solid |
| Chemical Name or Material | Pepstatin A |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?