missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pepsinogen I Antibody (8003 (99/12)) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB100-62239JF525
This item is not returnable.
View return policy
Description
Pepsinogen I Monoclonal antibody specifically detects Pepsinogen I in Human samples. It is validated for ELISA,Immunohistochemistry,Immunohistochemistry (Paraffin),Radioimmune Assays
Specifications
| Pepsinogen I | |
| Monoclonal | |
| Janelia Fluor 525 | |
| pepsinogen 3, group I (pepsinogen A) | |
| Purified human pepsinogen 1 | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 643834 | |
| Store at 4°C in the dark. | |
| IgG1 |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Radioimmune Assays (RIA) | |
| 8003 (99/12) | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion