missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pepsinogen A4/PGA4 Antibody (021), mFluor Violet 610 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Pepsinogen A4/PGA4 Monoclonal antibody specifically detects Pepsinogen A4/PGA4 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Pepsinogen A4/PGA4 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Dilution | Immunohistochemistry-Paraffin |
| Host Species | Rabbit |
| Content And Storage | Store at 4C in the dark. |
| Form | Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?