missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ PeproGMP® Human IL-3 Recombinant Protein, PeproTech®

Codice prodotto. 30276693 Sfoglia Tutto Gibco Prodotti
Cambia vista
Click to view available options
Quantity:
1 mg
100 μg
50 μg
Dimensione della confezione:
100µg
1mg
50µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity Dimensione della confezione
30276693 1 mg 1mg
30275259 100 μg 100µg
30275260 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30276693 Supplier Gibco™ Supplier No. GMP200031MG
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recombinant Protein

Additional product information and specifications are provided on the Certificate of Analysis. All PeproGMP® products are manufactured using animal-derived component free materials. Product Description/Intended Use: PeproTech GMP recombinant proteins are manufactured for use as ancillary materials by applying applicable principles of GMP and quality control requirements from USP (United States Pharmacopeia) Chapter 1043 Ancillary Materials for Cell, Gene, and Tissue-Engineered Products. Product use statement: For research use or further manufacturing. Not for diagnostic use or direct administration into humans or animals.

Interleukin 3 (IL-3), also called Mast Cell Growth Factor (MCGF), Multi-Colony Stimulating Factor (multi-CSF), or Eosinophil-CSF (E-CSF), is produced by activated T cells, mast cells and eosinophils. IL-3 is a hematopoietic growth factor that stimulates colony formation of erythroid, megakaryocyte, neutrophil, eosinophil, basophil, mast cell and monocytic lineages. Most of these functions are enhanced or dependent on co-stimulation with other cytokines. Human and mouse IL-3 share only 29% sequence identity at the amino acid level. The bioactivity of IL-3 is highly species specific. For example, human IL-3 has no activity on mouse cells. IL-3 is a potent growth promoting cytokine. IL-3 is capable of supporting the proliferation of a broad range of hematopoietic cell types, and is involved in a variety of cell activities such as cell growth, differentiation and apoptosis. IL-3 has been shown to also possess neurotrophic activity, and it may be associated with neurologic disorders.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P08700
For Use With (Application) Bioactivity, Control, Western Blot Control
Format Lyophilized
Formulation Protein with no preservative
Gene ID (Entrez) 3562
Molecular Weight (g/mol) 15 kDa
Name PeproGMP™ Human IL-3
Purification Method Purified
Quantity 1 mg
Storage Requirements -20°C or -80°C if preferred
Regulatory Status GMP
Endotoxin Concentration ≤0.1 EU/μg
Gene Alias BP/BPA; BPA; Colony stimulating factor multiple; colony-stimulating factor, multiple; Csfmu; Eo-CSF; HCGF; hematopoietic growth factor; Il3; Il-3; ILN; Interleukin; interleukin 3; Interleukin 3 (colony stimulating factor; Interleukin3; interleukin-3; Mast cell growth factor; mast-cell growth factor; MCGF; MGC79398; MGC79399; MGF; Multi CSF; MULTI-CSF; Multilineage colony stimulating factor; multilineage-colony-stimulating factor; multiple); multipotential colony-stimulating factor; P cell stimulating factor; P-cell stimulating factor; P-cell-stimulating factor; PCSA; PSF
Common Name IL-3
Gene Symbol IL3
Biological Activity Determined by the proliferation of TF-1 cells. PeproGMP Recombinant Human IL-3 ha been tested against the WHO standard NIBSC 91/510.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence APMTQTTSLK TSWVNCSNMI DEIITHLKQP PLPLLDFNNL NGEDQDILME NNLRRPNLEA FNRAVKSLQN ASAIESILKN LLPCLPLATA APTRHPIHIK DGDWNEFRRK LTFYLKTLEN AQAQQTTLSL AIF
Content And Storage -20°Cor to +80°C if preferred
Activity Determined by the proliferation of TF-1 cells. PeproGMP Recombinant Human IL-3 has been tested against the WHO standard NIBSC 91/510. Find the International Unit (IU) value <a href='https://www.thermofisher.com/us/en/home/life-science/cell-culture/cell-culture-learning-center/recombinant-protein-information/international-units.html'>here</a>.
Expression System E. coli
Form Lyophilized
Product Line PeproGMP
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.