missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ PeproGMP® Human IL-2 Recombinant Protein, PeproTech®

Product Code. 30276214
Change view
Click to view available options
Quantity:
1 mg
100 μg
50 μg
Unit Size:
100µg
1mg
50µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30276214 100 μg 100µg
30275501 1 mg 1mg
30275964 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
To receive the discount customers must purchase 2 of the same product at list price in a single order to receive 30% discount. There is no limit to the multiples of 2 that customers can buy. Use promo code ”27974” to get your promotional price
Product Code. 30276214 Supplier Gibco™ Supplier No. GMP20002100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase 2 of the same product at list price in a single order to receive 30% discount. There is no limit to the multiples of 2 that customers can buy. Use promo code ”27974” to get your promotional price

Recombinant Protein

Recombinant Human IL-2 is a 15.5 kDa protein, containing 134 amino acid residues including one intrachain disulfide bond. Additional product information and specifications are provided on the Certificate of Analysis. All PeproGMP® products are manufactured using animal-derived component free materials. Product Description/Intended Use: PeproTech GMP recombinant proteins are manufactured for use as ancillary materials by applying applicable principles of GMP and quality control requirements from USP (United States Pharmacopeia) Chapter 1043 Ancillary Materials for Cell, Gene, and Tissue-Engineered Products. Product use statement: For research use or further manufacturing. Not for diagnostic use or direct administration into humans or animals.

Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P60568
For Use With (Application) Control, Western Blot Control, Bioactivity
Format Lyophilized
Formulation Protein with no preservative
Gene ID (Entrez) 3558
Molecular Weight (g/mol) 15.5 kDa
Name PeproGMP™ Human IL-2
Purification Method Purified
Quantity 100 μg
Storage Requirements -20°C or -80°C if preferred
Regulatory Status GMP
Endotoxin Concentration ≤0.1 EU/μg
Gene Alias aldesleukin; cytokine; H-IL-2; il 2; Il2; Il-2; ILN; Interleukin; interleukin 2; interleukin 2 precursor; interleukin 2 variant IL2delta2; Interleukin2; Interleukin-2; interleukin-2 precursor; interleukin-2 precursor protein; interleukin-2; interleukin 2; Involved in regulation of T cell clonal expansion; involved in regulation of T-cell clonal expansion; lymphokine; M-IL-2; patial sequence of Interleukin-2; POIL2; porcine Interleukin 2; T cell growth factor; T-cell growth factor; T-cell growth factor (TCGF); TCGF
Common Name IL-2
Gene Symbol IL2
Biological Activity Determined by the proliferation of mouse CTLL-2 cells. PeproGMP Recombinant Human IL-2 ha been tested against the WHO standard NIBSC 86/500.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence MAPTSSSTKK TQLQLEHLLL DLQMILNGIN NYKNPKLTRM LTFKFYMPKK ATELKHLQCL EEELKPLEEV LNLAQSKNFH LRPRDLISNI NVIVLELKGS ETTFMCEYAD ETATIVEFLN RWITFAQSII STLT
Content And Storage -20°Cor to +80°C if preferred
Activity Determined by the proliferation of mouse CTLL-2 cells. PeproGMP Recombinant Human IL-2 has been tested against the WHO standard NIBSC 86/500. Find the International Unit (IU) value <a href='https://www.thermofisher.com/us/en/home/life-science/cell-culture/cell-culture-learning-center/recombinant-protein-information/international-units.html'>here</a>.
Expression System E. coli
Form Lyophilized
Product Line PeproGMP
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.