Learn More
Gibco™ PeproGMP™ Human FGF-basic (FGF-2/bFGF) Recombinant Protein, PeproTech®
Description
The PeproTech Recombinant Human FGF-basic is a 17.2 kDa protein consisting of 154 amino acid residues. Reconstitute using sterile water (i.e., WFI) to a concentration of 0.1-1.0 mg/ml. Additional product information, specifications, handling and reconstitution information are provided on the Certificate of Analysis. All PeproGMP™ products are manufactured using animal-derived component free materials. Product Description/Intended Use: PeproTech GMP recombinant proteins are manufactured for use as ancillary materials by applying applicable principles of GMP and quality control requirements from USP (United States Pharmacopeia) Chapter <1043> Ancillary Materials for Cell, Gene, and Tissue-Engineered Products.
Specifications
Specifications
| Accession Number | P09038 |
| For Use With (Application) | Control, Western Blot Control, Bioactivity |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 2247 |
| Molecular Weight (g/mol) | 17.2 kDa |
| Name | PeproGMP™ Human FGF-basic (FGF-2/bFGF) |
| Purification Method | Purified |
| Quantity | 25 μg |
| Storage Requirements | -20°C or -80°C if preferred |
| Regulatory Status | GMP |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.