missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEPP2 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-37696C
This item is not returnable.
View return policy
Description
PEPP2 Polyclonal antibody specifically detects PEPP2 in Human samples. It is validated for Western Blot
Specifications
| PEPP2 | |
| Polyclonal | |
| Western Blot | |
| FLJ10667, FLJ26734, KIAA1686FLJ31492, PEPP-2, PEPP2phosphoinositol 3-phosphate-binding protein-2, PH domain-containing family A member 5, Phosphoinositol 3-phosphate-binding protein 2, pleckstrin homology domain containing, family A member 5, pleckstrin homology domain-containing family A member 5 | |
| This antibody maps to a region between residue 876 to 925 of human Pleckstrin Homology Domain Containing, Family A Member 6 using the numbering given in entry NP_061885.3 (GeneID 54477). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 54477 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction