Learn More
Description
Separate hydrophobic peptides and proteins, providing higher recoveries with Thermo Scientific™ Acclaim™ PepMap™ 300 C4 LC Columns.
- High resolution in protein identification, biomarker discovery and systems biology
- Particular loadability allows hghest sensitivity in LC/MS
- Designed for TFA-free LC-MS, minimizing ion-suppression effects
- Optimized for coupling to ESI/MS and MALDI-MS
- Highest column-to-column reproducibility
- Easy-to-use, cutting-edge miniaturized HPLC
Spécification
Spécification
| Column Format | Analytical Column |
| Diameter (Metric) | 0.075 mm |
| Flow Rate | 200 nL/min |
| For Use With | LC/MS, ESI-MS, MALDI-MS |
| Length (Metric) | 150 mm |
| Max. Pressure | 7250 psi (500 bar) |
| Particle Size | 5 μm |
| pH | 2 to 8 |
| Pore Size | 300 Å |
| Surface Area | 450 m²/g |
| Afficher plus |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.