missing translation for 'onlineSavingsMsg'
Learn More

PEPD Antibody (OTI5D5) [DyLight 405], Novus Biologicals™

Product Code. 30687681 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30687681 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30687681 Supplier Novus Biologicals Supplier No. NBP344638V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PEPD Monoclonal antibody specifically detects PEPD in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEPD
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI5D5
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias aminoacyl-L-proline hydrolase, EC 3.4.13.9, Imidodipeptidase, MGC10905, peptidase Dimidodipeptidase, PRD, PROLIDASE, Proline dipeptidase, xaa-Pro dipeptidase, X-Pro dipeptidase
Host Species Mouse
Immunogen Full length human recombinant protein of human PEPD (NP_000276) produced in HEK293T cell.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 5184
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.