missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PEO1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551715
This item is not returnable.
View return policy
Description
Immunogen sequence: SKAPEFEDSE EVRRIWNRAI PLWELPDQEE VQLADTMFGL TKVTDDTLKR FSVRYLRPAR SLVFPWFSPG GSGLRGLKLL EAKCQGDGVS YEETTIPRPS AYHNLFGLPL ISRRDAEVVL TSRELDSLAL NQSTG Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 78%.
Twinkle is a mitochondrial protein with structural similarity to the phage T7 primase/helicase (GP4) and other hexameric ring helicases. The twinkle protein colocalizes with mtDNA in mitochondrial nucleoids, and its name derives from the unusual localization pattern reminiscent of twinkling stars.Twinkle is a mitochondrial protein with structural similarity to the phage T7 primase/helicase (GP4) and other hexameric ring helicases. The twinkle protein colocalizes with mtDNA in mitochondrial nucleoids, and its name derives from the unusual localization pattern reminiscent of twinkling stars (Spelbrink et al., 2001 [PubMed 11431692]).[supplied by OMIM]. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.
Specifications
| PEO1 | |
| Polyclonal | |
| Unconjugated | |
| TWNK | |
| ataxin 8; Ataxin8; ATXN 8; ATXN8; C10 orf2; C10orf 2; C10orf2; C18H10orf2; C1H10orf2; C26H10orf2; C28H10orf2; chromosome 10 open reading frame 2; D19Ertd626e; fi04d09; FLJ21832; IOSCA; mitochondrial twinkle protein; MTDPS7; PEO; PEO 1; peo1; PEOA3; PRLTS5; progressive external ophthalmoplegia 1; progressive external ophthalmoplegia 1 (human); progressive external ophthalmoplegia 1 homolog; progressive external ophthalmoplegia 1 protein; Progressive external ophthalmoplegia 1 protein homolog; SANDO; SCA 8; SCA8; T7 gp4-like protein with intramitochondrial nucleoid localization; T7 helicase-related protein with intramitochondrial nucleoid localization; T7-like mitochondrial DNA helicase; twinkle; Twinkle mtDNA helicase; twinkle protein, mitochondrial; twinkle protein, mitochondrial-like; Twinkle protein, mitochondrial-like protein; Twinl; Twnk; wu:fi04d09 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 56652 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96RR1 | |
| TWNK | |
| Recombinant protein corresponding to Human PEO1. Recombinant protein control fragment (Product #RP-102228). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction