missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PEO1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541289
This item is not returnable.
View return policy
Description
Peptide sequence: GVFRKFATDN NCHVTLVIHP RKEDDDKELQ TASIFGSAKA SQEADNVLIL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
Twinkle is a mitochondrial protein with structural similarity to the phage T7 primase/helicase (GP4) and other hexameric ring helicases. The twinkle protein colocalizes with mtDNA in mitochondrial nucleoids, and its name derives from the unusual localization pattern reminiscent of twinkling stars.Twinkle is a mitochondrial protein with structural similarity to the phage T7 primase/helicase (GP4) and other hexameric ring helicases. The twinkle protein colocalizes with mtDNA in mitochondrial nucleoids, and its name derives from the unusual localization pattern reminiscent of twinkling stars (Spelbrink et al., 2001 [PubMed 11431692]).[supplied by OMIM]. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.
Specifications
| PEO1 | |
| Polyclonal | |
| Unconjugated | |
| TWNK | |
| ataxin 8; Ataxin8; ATXN 8; ATXN8; C10 orf2; C10orf 2; C10orf2; C18H10orf2; C1H10orf2; C26H10orf2; C28H10orf2; chromosome 10 open reading frame 2; D19Ertd626e; fi04d09; FLJ21832; IOSCA; mitochondrial twinkle protein; MTDPS7; PEO; PEO 1; peo1; PEOA3; PRLTS5; progressive external ophthalmoplegia 1; progressive external ophthalmoplegia 1 (human); progressive external ophthalmoplegia 1 homolog; progressive external ophthalmoplegia 1 protein; Progressive external ophthalmoplegia 1 protein homolog; SANDO; SCA 8; SCA8; T7 gp4-like protein with intramitochondrial nucleoid localization; T7 helicase-related protein with intramitochondrial nucleoid localization; T7-like mitochondrial DNA helicase; twinkle; Twinkle mtDNA helicase; twinkle protein, mitochondrial; twinkle protein, mitochondrial-like; Twinkle protein, mitochondrial-like protein; Twinl; Twnk; wu:fi04d09 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 56652 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96RR1 | |
| TWNK | |
| Synthetic peptide directed towards the middle region of human PEO1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction