missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PEO1 Monoclonal antibody specifically detects PEO1 in Human samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | PEO1 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 1C5 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | NP_068602 |
| Gene Alias | ATXN8, chromosome 10 open reading frame 2, EC 3.6.4.12, FLJ21832, infantile onset spinocerebellar ataxia (autosomal recessive), IOSCA, mitochondrial, MTDPS7, PEO1PEOA3, TWINKLE |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?