missing translation for 'onlineSavingsMsg'
Learn More

PEO-IAA, MedChemExpress

Product Code. 16400987
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16400987

Brand: Medchem Express HY112730

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PEO-IAA is an indole-3-acetic acid (IAA) antagonist. PEO-IAA is an auxin antagonist that binds to transport inhibitor response 1/auxin signaling F-box proteins (TIR1/AFBs).

PEO-IAA is an indole-3-acetic acid (IAA) antagonist. PEO-IAA is an auxin antagonist that binds to transport inhibitor response 1/auxin signaling F-box proteins (TIR1/AFBs).

Specifications

Chemical Name or Material PEO-IAA
Physical Form Solid
CAS 6266-66-6
Color Light Nude
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C18H15NO3
Synonym 2-(1H-Indol-3-yl)-4-oxo-4-phenyl-butyric acid
Solubility Information DMSO : 50 mg/mL (170.46 mM; Need ultrasonic)
SMILES O=C(O)C(CC(C1=CC=CC=C1)=O)C2=CNC3=C2C=CC=C3
Molecular Weight (g/mol) 293.32
Formula Weight 293.32
Percent Purity 98.08%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.