missing translation for 'onlineSavingsMsg'
Learn More

Pentylmagnesium bromide, 2M solution in diethyl ether, AcroSeal™

Product Code. p-4228863 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
This item is not returnable. View return policy

Product Code. 11935961

Brand: Thermo Scientific Acros 435551000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 693-25-4 | C5H11BrMg | 175.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 693-25-4, 60-29-7
Molecular Formula C5H11BrMg
Molecular Weight (g/mol) 175.35
MDL Number MFCD00000045
InChI Key ZXWGTPYWPZGZPT-UHFFFAOYSA-M
Synonym amylmagnesium bromide, magnesium, bromopentyl, bromopentylmagnesium, n-pentylmagnesium bromide, pentylmagnesium bromide, pentylmagnesium bromide solution, n-pentyl magnesium bromide, npentmgbr;, pentylmagnesiumbromide, amyl-magnesium bromide, grignard reagent
PubChem CID 101838
IUPAC Name magnesium;pentane;bromide
SMILES CCCCC[Mg]Br

Specifications

Color Black to Yellow
Density 0.9470g/mL
Flash Point -40°C
Packaging AcroSeal™ Glass Bottle
Quantity 100 mL
Specific Gravity 0.947
Solubility Information Solubility in water: reacts
Formula Weight 175.35
Physical Form Solution
Chemical Name or Material Pentylmagnesium bromide
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Pentylmagnesium bromide, 2M solution in diethyl ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H224-Extremely flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P243-Take precautionary measures against static discharge.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.