missing translation for 'onlineSavingsMsg'
Learn More

Pentyl Paraben, TRC

Product Code. 30132403
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132403 1 g 1g
17942735 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30132403 Supplier Toronto Research Chemicals Supplier No. TRCP2838001G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6521-29-5
Molecular Formula C12 H16 O3
Molecular Weight (g/mol) 208.25
Synonym Benzoic acid, 4-hydroxy-, pentyl ester, Benzoic acid, p-hydroxy-, pentyl ester (6CI,8CI), 4-Hydroxybenzoic acid n-amyl ester, Amyl p-hydroxybenzoate, NSC 309817, Pentyl 4-hydroxybenzoate, Pentyl p-hydroxybenzoate, Pentyl paraben, n-Amyl 4-hydroxybenzoate, 4-Hydroxybenzoic acid pentyl ester
IUPAC Name pentyl 4-hydroxybenzoate
SMILES CCCCCOC(=O)c1ccc(O)cc1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H16O3/c1-2-3-4-9-15-12(14)10-5-7-11(13)8-6-10/h5-8,13H,2-4,9H2,1H3
Formula Weight 208.1099
Chemical Name or Material Pentyl Paraben
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.