missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentraxin 3/TSG-14 Antibody (MM0512-6B7), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-11849CL1
This item is not returnable.
View return policy
Description
Pentraxin 3/TSG-14 Monoclonal antibody specifically detects Pentraxin 3/TSG-14 in Human samples. It is validated for Western Blot
Specifications
| Pentraxin 3/TSG-14 | |
| Monoclonal | |
| CoraFluor 1 | |
| alpha-induced protein 5, pentaxin-related gene, rapidly induced by IL-1 beta, tumor necrosis factor, Pentaxin-related protein PTX3, pentraxin 3, long, pentraxin-3, pentraxin-related gene, rapidly induced by IL-1 beta, pentraxin-related protein PTX3, TNF alpha-induced protein 5, TNFAIP5, TSG14, TSG-14pentaxin-related gene, rapidly induced by IL-1 beta, Tumor necrosis factor alpha-induced protein 5, tumor necrosis factor, alpha-induced protein 5, Tumor necrosis factor-inducible gene 14 protein, tumor necrosis factor-inducible protein TSG-14 | |
| Human recombinant Pentraxin 3/TSG-14 | |
| 0.1 mL | |
| Immunology | |
| 5806 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2 |
| Western Blot | |
| MM0512-6B7 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction