missing translation for 'onlineSavingsMsg'
Learn More

Pentoxyverine Hydrogen Citrate, Mikromol

Product Code. 30461315
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30461315 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30461315 Supplier Mikromol Supplier No. MM1268.000250
Big Savings for Better Separation
Use promo code 27992
View all promotions
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Pentoxyverine Hydrogen Citrate, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pentoxyverine Citrate
CAS 23142-01-0
Analyte or Component Names Pentoxyverine Citrate
Shipping Condition Room Temperature
Molecular Formula C20 H31 N O3 . C6 H8 O7
Certifications/Compliance ISO 17034
Synonym Pentoxyverine hydrogen citrate, 2-[2-(Diethylamino)ethoxy]ethyl 1-phenylcyclopentanecarboxylate dihydrogen 2-hydroxypropane-1,2,3-tricarboxylate, Cyclopentanecarboxylic acid, 1-phenyl-, 2-[2-(diethylamino)ethoxy]ethyl ester, 2-hydroxy-1,2,3-propanetricarboxylate (1:1), Cyclopentanecarboxylic acid, 1-phenyl-, 2-[2-(diethylamino)ethoxy]ethyl ester citrate (1:1) (8CI), Antees, Calnathal, Carbetane, Carbetapentane citrate, Cossym, Fustpentane, Germapect, Pencal, Pentoxyverine citrate, Sedotussin, Toclase, Tosnore, Tuclase, UCB 2543
InChI Formula InChI=1S/C20H31NO3.C6H8O7/c1-3-21(4-2)14-15-23-16-17-24-19(22)20(12-8-9-13-20)18-10-6-5-7-11-18;7-3(8)1-6(13,5(11)12)2-4(9)10/h5-7,10-11H,3-4,8-9,12-17H2,1-2H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)
Solution Type Neat
SMILES CCN(CC)CCOCCOC(=O)C1(CCCC1)c2ccccc2.OC(=O)CC(O)(CC(=O)O)C(=O)O
IUPAC Name 2-[2-(diethylamino)ethoxy]ethyl 1-phenylcyclopentane-1-carboxylate;2-hydroxypropane-1,2,3-tricarboxylic acid
Molecular Weight (g/mol) 525.59
Quantity 250 mg
Formula Weight 525.2574 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.