missing translation for 'onlineSavingsMsg'
Learn More

Pentoxyverine Citrate, TRC

Product Code. 17932655
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17932655 5 g 5g
17911545 100 mg 100mg
17903895 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17932655 Supplier Toronto Research Chemicals Supplier No. TRCP2766005G
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23142-01-0
Molecular Formula C20 H31 N O3 . C6 H8 O7
Molecular Weight (g/mol) 525.59
Synonym Pentoxyverine hydrogen citrate, 2-[2-(Diethylamino)ethoxy]ethyl 1-phenylcyclopentanecarboxylate dihydrogen 2-hydroxypropane-1,2,3-tricarboxylate, Cyclopentanecarboxylic acid, 1-phenyl-, 2-[2-(diethylamino)ethoxy]ethyl ester, 2-hydroxy-1,2,3-propanetricarboxylate (1:1), Cyclopentanecarboxylic acid, 1-phenyl-, 2-[2-(diethylamino)ethoxy]ethyl ester citrate (1:1) (8CI), Antees, Calnathal, Carbetane, Carbetapentane citrate, Cossym, Fustpentane, Germapect, Pencal, Pentoxyverine citrate, Sedotussin, Toclase, Tosnore, Tuclase, UCB 2543
IUPAC Name 2-[2-(diethylamino)ethoxy]ethyl 1-phenylcyclopentane-1-carboxylate;2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES CCN(CC)CCOCCOC(=O)C1(CCCC1)c2ccccc2.OC(=O)CC(O)(CC(=O)O)C(=O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C20H31NO3.C6H8O7/c1-3-21(4-2)14-15-23-16-17-24-19(22)20(12-8-9-13-20)18-10-6-5-7-11-18;7-3(8)1-6(13,5(11)12)2-4(9)10/h5-7,10-11H,3-4,8-9,12-17H2,1-2H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)
Formula Weight 525.2574
Percent Purity >95%
Chemical Name or Material Pentoxyverine Citrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.