missing translation for 'onlineSavingsMsg'
Learn More

Pentoxifylline-4',4',6',6',6'-d5, TRC

Product Code. 30419507
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
Unit Size:
0.5mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30419507 1 mg 1mg
30415234 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30419507 Supplier Toronto Research Chemicals Supplier No. TRCP2765011MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pentoxifylline-4',4',6',6',6'-d5
CAS 1185995-18-9
Molecular Formula C13 D5 H13 N4 O3
Quantity 1 mg
Synonym 1H-Purine-2,6-dione','3,7-dihydro-3,7-dimethyl-1-(5-oxohexyl-4,4,6,6,6-d5)- (ACI), 3,7-Dihydro-3,7-dimethyl-1-(5-oxohexyl-4,4,6,6,6-d5)-1H-purine-2,6-dione (ACI), C 21256
InChI Formula InChI=1S/C13H18N4O3/c1-9(18)6-4-5-7-17-12(19)10-11(14-8-15(10)2)16(3)13(17)20/h8H,4-7H2,1-3H3/i1D3,6D2
SMILES [2H]C([2H])([2H])C(=O)C([2H])([2H])CCCN1C(=O)N(C)c2ncn(C)c2C1=O
IUPAC Name 3,7-dimethyl-1-(4,4,6,6,6-pentadeuterio-5-oxohexyl)purine-2,6-dione
Molecular Weight (g/mol) 283.34
Formula Weight 283.1693
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.