missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentiapine, MedChemExpress
Description
Pentiapine (CGS 10746) is a dopamine release inhibitor without binding to synaptic dopamine receptor sites.
Specifications
Specifications
| Chemical Name or Material | Pentiapine |
| CAS | 81382-51-6 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C15H17N5S |
| Quantity | 5 mg |
| Synonym | CGS 10746 |
| Solubility Information | DMSO : 100 mg/mL (334.01 mM; Need ultrasonic) |
| SMILES | CN1CCN(C2=NC3=CC=CC=C3SC4=NC=CN42)CC1 |
| Molecular Weight (g/mol) | 299.39 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?