Learn More
Pentel™ Pentel superfine marker, black
Product Code. 15432554
Encompass_Preferred
Product Code. 15432554
Brand: Pentel™ 8350596
This item requires a price update prior to being ordered.
Request a Price for this item here.
Pentel MF50 is a super fine permanent marker ideal for intricate marking or labelling. Xylene and toluene free with a durable plastic tip producing a 0.4mm line width.
Black