missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentanol, mixture of isomers, 99% (sum of isomers)
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 0.815 |
| Boiling Point | 128°C to 137°C |
| Flash Point | 45°C (113°F) |
| Refractive Index | 1.4102 |
| Quantity | 1000 mL |
| UN Number | UN1987 |
| Formula Weight | 88.15 |
| Percent Purity | 99% |
| Chemical Name or Material | Pentanol, mixture of isomers |
Safety and Handling
Product Identifier
- Pentanol, mixture of isomers
- Warning
- Acute toxicity Category 4
- Flammable liquid Category 3
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- MIXTURE LIST-Contain: Pentanol, branched and linear
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?