missing translation for 'onlineSavingsMsg'
Learn More

Pentanol, mixture of isomers, 99% (sum of isomers)

Product Code. 15408445
Change view
Click to view available options
Quantity:
500 mL
1000 mL
5000 mL
Unit Size:
1000mL
5000mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15408445 500 mL 500mL
15418445 1000 mL 1000mL
15498435 5000 mL 5000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15408445 Supplier Thermo Scientific Alfa Aesar Supplier No. H33391.AP
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 94624-12-1 | C5H12O | 88.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94624-12-1
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00081734
InChI Key AMQJEAYHLZJPGS-UHFFFAOYSA-N
Synonym 1-pentanol, amyl alcohol, n-amyl alcohol, n-pentanol, pentanol, pentyl alcohol, butylcarbinol, 1-pentyl alcohol, amylol, n-butylcarbinol
PubChem CID 6276
ChEBI CHEBI:44884
IUPAC Name pentan-1-ol
SMILES CCCCCO

Specifications

Density 0.815
Boiling Point 128°C to 137°C
Flash Point 45°C (113°F)
Refractive Index 1.4102
Quantity 500 mL
UN Number UN1987
Formula Weight 88.15
Percent Purity 99%
Chemical Name or Material Pentanol, mixture of isomers

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.