missing translation for 'onlineSavingsMsg'
Learn More

Pentanoic-5,5,5-d3 Acid, TRC

Product Code. 30422644
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30422644 50 mg
30421322 100 mg
30421262 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30422644 Supplier Toronto Research Chemicals Supplier No. TRCP27436050MG
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83741-76-8
Molecular Formula C5 D3 H7 O2
Molecular Weight (g/mol) 105.15
Synonym Pentanoic-5,5,5-d3 acid (9CI), δ-Trideuteropentanoic acid, Pentanoic acid-5,5,5-d3, Valeric acid-d3, Valeric-δ,δ,δ-d3 acid, Pentanoic-5,5,5-D3 acid
IUPAC Name 5,5,5-trideuteriopentanoic acid
SMILES [2H]C([2H])([2H])CCCC(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C5H10O2/c1-2-3-4-5(6)7/h2-4H2,1H3,(H,6,7)/i1D3
Formula Weight 105.087
Chemical Name or Material Pentanoic-5,5,5-d3 Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.