missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentanenitrile-d9, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.10g
0.25g
Specifications
Specifications
| Chemical Name or Material | Pentanenitrile-d9 |
| CAS | 1030939-70-8 |
| Molecular Formula | C5 D9 N |
| Quantity | 0.25 g |
| Synonym | Pentanenitrile-2,2,3,3,4,4,5,5,5-d9 (ACI), Perdeuterated-1-cyanobutane |
| InChI Formula | InChI=1 S/C5H9N/c1-2-3-4-5-6/h2-4H2,1H3/i1D3,2D2,3D2,4D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C#N |
| IUPAC Name | 2,2,3,3,4,4,5,5,5-nonadeuteriopentanenitrile |
| Molecular Weight (g/mol) | 92.19 |
| Formula Weight | 92.13 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction