missing translation for 'onlineSavingsMsg'
Learn More

Pentanehydrazide, TRC

Product Code. 30428705
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30428705 10 mg 10mg
30416999 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30428705 Supplier Toronto Research Chemicals Supplier No. TRCP27404810MG
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pentanehydrazide
CAS 38291-82-6
Molecular Formula C5 H12 N2 O
Quantity 10 mg
InChI Formula InChI=1S/C5H12N2O/c1-2-3-4-5(8)7-6/h2-4,6H2,1H3,(H,7,8)
SMILES CCCCC(=O)NN
IUPAC Name pentanehydrazide
Molecular Weight (g/mol) 116.162
Formula Weight 116.095
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.