missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentane, for HPLC, Mixed Isomers, Fisher Chemical™
£52.65 - £127.00
Chemical Identifiers
CAS | 109-66-0 |
---|---|
Molecular Formula | C5H12 |
Molecular Weight (g/mol) | 72.15 |
MDL Number | MFCD00009498 |
InChI Key | OFBQJSOFQDEBGM-UHFFFAOYSA-N |
Synonym | n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish |
PubChem CID | 8003 |
ChEBI | CHEBI:37830 |
IUPAC Name | pentane |
SMILES | CCCCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10060280
|
Fisher Chemical
P/1006/15 |
1 L |
£52.65
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10346000
|
Fisher Chemical
P/1006/17 |
2.5 L |
£127.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
109-66-0 | |
72.15 | |
OFBQJSOFQDEBGM-UHFFFAOYSA-N | |
8003 | |
pentane |
C5H12 | |
MFCD00009498 | |
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish | |
CHEBI:37830 | |
CCCCC |
Specifications
-130°C | |
36°C | |
Liquid | |
MFCD00009498 | |
0.25 mPaS at 20°C | |
OFBQJSOFQDEBGM-UHFFFAOYSA-N | |
pentane | |
8003 | |
72.15g/mol |
109-66-0 | |
Amber Glass Bottle | |
C5H12 | |
573mbar at 20°C | |
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish | |
CCCCC | |
72.15 | |
CHEBI:37830 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Pentane mixed isomers
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H336-May cause drowsiness or dizziness.
- H411-Toxic to aquatic life with long lasting effects.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P273-Avoid release to the environment.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.