missing translation for 'onlineSavingsMsg'
Learn More

Pentan-1-ol, Certified AR for Analysis, Fisher Chemical™

Product Code. 11408453 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Unit Size:
2.5 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11408453 500 mL 500mL
10568440 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11408453 Supplier Fisher Chemical Supplier No. P/1041/PB08
  • ONLINE EXCLUSIVE PRICE
    £117.83
    £84.20 / 500mL
    Save £33.63
    28% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 71-41-0
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00002977,MFCD00081734,MFCD01075169
InChI Key AMQJEAYHLZJPGS-UHFFFAOYSA-N
Synonym 1-pentanol, amyl alcohol, n-amyl alcohol, n-pentanol, pentanol, pentyl alcohol, butylcarbinol, 1-pentyl alcohol, amylol, n-butylcarbinol
PubChem CID 6276
ChEBI CHEBI:44884
IUPAC Name pentan-1-ol
SMILES CCCCCO

Specifications

Melting Point -78°C
Color Colorless
pH 7
Boiling Point 137°C
Packaging Plastic Coated Glass bottle
Quantity 500 mL
Vapor Density 3
Formula Weight 88.15g/mol
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • 1-Pentanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H332-Harmful if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.