missing translation for 'onlineSavingsMsg'
Learn More

Pentamethylenebis(magnesium bromide), 0.5M solution in THF, AcroSeal™

Product Code. 11965971 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11965971 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11965971 Supplier Thermo Scientific Acros Supplier No. 436001000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 23708-48-7 | C5H10Br2Mg2 | 278.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 23708-48-7, 109-99-9
Color Brown to Yellow or Gray
Density 0.9990g/mL
Flash Point −17°C
Packaging AcroSeal™ Glass Bottle
Molecular Formula C5H10Br2Mg2
MDL Number MFCD00012384
Quantity 100 mL
Specific Gravity 0.999
Synonym pentamethylenebis magnesium bromide, bromo 5-bromomagnesio pentyl magnesium, pentane-1,5-di magnesium bromide, pentamethylenebis magnesium bromide solution, pentamethylenebis magnesium bromide 0,5m w/w in thf, pentamethylenebis magnesium bromide solution, 0.5 m in thf, grignard reagent
Solubility Information Solubility in water: reacts.
InChI Key OUEGHZYBWJTUAD-UHFFFAOYSA-L
SMILES Br[Mg]CCCCC[Mg]Br
IUPAC Name dimagnesium;pentane;dibromide
Molecular Weight (g/mol) 278.55
PubChem CID 3637391
Formula Weight 278.55
Physical Form Solution
Chemical Name or Material Pentamethylenebis(magnesium bromide)
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Pentamethylenebis(magnesium bromide), 0.5M solution in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which, in contact with water, emits flammable gases Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H261-In contact with water releases flammable gases.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.