missing translation for 'onlineSavingsMsg'
Learn More

Pentamethylcyclopentadienylzirconium trichloride, 99%, Thermo Scientific Chemicals

£113.00 - £335.00

Chemical Identifiers

CAS 75181-07-6
MDL Number MFCD00070444
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11348358
View Documents Promotion Details
Thermo Scientific Alfa Aesar
039551.03
1 g
£113.00
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11338358
View Documents Promotion Details
Thermo Scientific Alfa Aesar
039551.06
5 g
£335.00
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

75181-07-6
MFCD00070444

Specifications

75181-07-6
Ampule
Moisture sensitive
99%
Pentamethylcyclopentadienylzirconium trichloride
230°C (decomposition)
MFCD00070444
332.81
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only