missing translation for 'onlineSavingsMsg'
Learn More

Pentamethylcyclopentadienyltitanium trichloride, Thermo Scientific Chemicals

£390.00

Chemical Identifiers

CAS 12129-06-5
MDL Number MFCD00070434
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11354517
View Documents Promotion Details
Thermo Scientific Alfa Aesar
039596.06
5 g
£390.00
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used as a catalyst in Transesterification reactions and styrene polymerization. It is also used as Piano-stool catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12129-06-5
MFCD00070434

Specifications

12129-06-5
Red
UN3261
Reacts with water
Crystalline
200°C (decomposition)
MFCD00070434
Moisture sensitive
289.47
Pentamethylcyclopentadienyltitanium trichloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only