missing translation for 'onlineSavingsMsg'
Learn More

Pentamethylcyclopentadienylmolybdenum dicarbonyl dimer, 99%

Product Code. 11373708
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11373708 1 g 1g
11383708 250 mg 250mg
11363708 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11373708 Supplier Thermo Scientific Alfa Aesar Supplier No. 039303.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12132-04-6 | C12H16MoO2 | 288.208 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12132-04-6
Molecular Formula C12H16MoO2
Molecular Weight (g/mol) 288.208
MDL Number MFCD00058846
InChI Key CIKWUWWMMYQFKN-UHFFFAOYSA-N
Synonym Dicarbonyl(pentamethylcyclopentadienyl)molybdenum dimer
PubChem CID 131675733
IUPAC Name carbon monoxide;molybdenum;1,2,3,4,5-pentamethylcyclopenta-1,3-diene
SMILES CC1C(=C(C(=C1C)C)C)C.[C-]#[O+].[C-]#[O+].[Mo]

Specifications

Melting Point 219°C (decomposition)
Quantity 1 g
UN Number UN3466
Formula Weight 574.38
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material Pentamethylcyclopentadienylmolybdenum dicarbonyl dimer

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.