missing translation for 'onlineSavingsMsg'
Learn More

Pentamethylcyclopentadienyliron dicarbonyl dimer

Product Code. 11333668
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11333668 5 g 5g
11343668 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11333668 Supplier Thermo Scientific Alfa Aesar Supplier No. 039266.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 35344-11-7 | C24H30Fe2O4 | 494.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35344-11-7
Molecular Formula C24H30Fe2O4
Molecular Weight (g/mol) 494.19
MDL Number MFCD00058862
InChI Key UXGBSDPRTITVSY-UHFFFAOYSA-N
Synonym pentamethylcyclopentadienyliron dicarbonyl dimer, carbon monoxide; iron; 1,2,3,4,5-pentamethylcyclopentane
PubChem CID 498645
IUPAC Name carbon monoxide;iron;1,2,3,4,5-pentamethylcyclopentane
SMILES [Fe].[Fe].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].C[C]1[C](C)[C](C)[C](C)[C]1C.C[C]1[C](C)[C](C)[C](C)[C]1C

Specifications

Color Purple-Red
Quantity 5 g
UN Number UN3466
Formula Weight 493.96
Physical Form Crystalline
Chemical Name or Material Pentamethylcyclopentadienyliron dicarbonyl dimer

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.