missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorothiophenol, TRC

Product Code. 30136098
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136098 50 mg 50mg
30132553 100 mg 100mg
30007694 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136098 Supplier Toronto Research Chemicals Supplier No. TRCP27356550MG
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 771-62-0
Molecular Formula C6HF5S
Molecular Weight (g/mol) 200.12
IUPAC Name 2,3,4,5,6-pentafluorobenzenethiol
SMILES Fc1c(F)c(F)c(S)c(F)c1F

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C6HF5S/c7-1-2(8)4(10)6(12)5(11)3(1)9/h12H
Formula Weight 199.9719
Chemical Name or Material Pentafluorothiophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.