missing translation for 'onlineSavingsMsg'
Learn More

Pentafluoropropionic acid, 97%

Product Code. 11435143
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11435143 25 g 25g
11445143 100 g 100g
11455143 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11435143 Supplier Thermo Scientific Alfa Aesar Supplier No. A12791.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 422-64-0 | C3HF5O2 | 164.03 g/mol

Pentafluoropropionic acid is used as an efficient catalyst for the preparation of dibenzo[a, j] xanthenes by condensation reaction of beta-naphthol with aryl aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentafluoropropionic acid is used as an efficient catalyst for the preparation of dibenzo[a, j] xanthenes by condensation reaction of beta-naphthol with aryl aldehydes.

Notes
Hygroscopic. Incompatible withstrong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 422-64-0
Molecular Formula C3AgF5O2
Molecular Weight (g/mol) 270.89
MDL Number MFCD00004170
InChI Key XAYJXAUUXJTOSI-UHFFFAOYSA-M
Synonym pentafluoropropionic acid, perfluoropropionic acid, pentafluoropropanoic acid, propanoic acid, pentafluoro, propionic acid, pentafluoro, 2,2,3,3,3-pentafluoropropionic acid, ipc-pffa-3, perfluoropropanoic acid, propanoic acid, 2,2,3,3,3-pentafluoro, perfluoropropionicacid
PubChem CID 62356
IUPAC Name 2,2,3,3,3-pentafluoropropanoic acid
SMILES [Ag+].[O-]C(=O)C(F)(F)C(F)(F)F

Specifications

Density 1.576
Boiling Point 95°C to 97°C
Odor Rancid
Refractive Index 1.284
Quantity 25 g
UN Number UN3265
Beilstein 1773387
Sensitivity Hygroscopic
Formula Weight 164.03
Percent Purity 97%
Chemical Name or Material Pentafluoropropionic acid
Show More Show Less
compliance-icons
Product Identifier
  • Pentafluoropropionic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Pentafluoropropionicacid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.