missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl trifluoroacetate, 98+%

Product Code. 15450808
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15450808 25 g 25g
15440808 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15450808 Supplier Thermo Scientific Alfa Aesar Supplier No. H54529.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14533-84-7 | C8F8O2 | 280.07 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Soluble in water at 122.5°Cat760mmHg.

Notes
Hygroscopic. Store at room temperature. Avoid strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14533-84-7
Molecular Formula C8F8O2
Molecular Weight (g/mol) 280.07
MDL Number MFCD00134438
InChI Key VCQURUZYYSOUHP-UHFFFAOYSA-N
Synonym pentafluorophenyl trifluoroacetate, perfluorophenyl 2,2,2-trifluoroacetate, trifluoroacetic acid pentafluorophenyl ester, pentafluorphenyl trifluoracetate, pentafluorophenyltrifluoroacetate, pentafluorophenyl 2,2,2-trifluoroacetate, acetic acid, trifluoro-, pentafluorophenyl ester, acetic acid,2,2,2-trifluoro-, 2,3,4,5,6-pentafluorophenyl ester, ambotzrl-1046, acmc-1bui8
PubChem CID 4327891
IUPAC Name (2,3,4,5,6-pentafluorophenyl) 2,2,2-trifluoroacetate
SMILES FC1=C(F)C(F)=C(OC(=O)C(F)(F)F)C(F)=C1F

Specifications

Density 1.63
Boiling Point 122°C to 123°C
Flash Point 52°C (126°F)
Refractive Index 1.368
Quantity 25 g
UN Number UN3272
Beilstein 2003848
Sensitivity Moisture sensitive
Solubility Information Soluble in water at 122.5°Cat760mmHg.
Formula Weight 280.07
Percent Purity ≥98%
Chemical Name or Material Pentafluorophenyl trifluoroacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.