missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl 4-(1-methyl-1H-pyrazol-3-yl)benzoate, 97%, Thermo Scientific™

Product Code. p-3541157
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11360587 250 mg Amber Glass Bottle 250mg
10472844 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11360587 Supplier Thermo Scientific Maybridge Supplier No. CC23826CB
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 915707-42-5
Molecular Formula C17H9F5N2O2
Molecular Weight (g/mol) 368.26
MDL Number MFCD09702352
InChI Key LGHZGIHYFYLEAC-UHFFFAOYSA-N
Synonym pentafluorophenyl 4-1-methyl-1h-pyrazol-3-yl benzoate, 2,3,4,5,6-pentafluorophenyl 4-1-methylpyrazol-3-yl benzoate, pentafluorophenyl 4-1-methylpyrazol-3-yl benzoate, perfluorophenyl 4-1-methyl-1h-pyrazol-3-yl benzoate, 2,3,4,5,6-pentakis fluoranyl phenyl 4-1-methylpyrazol-3-yl benzoate, 4-1-methyl-3-pyrazolyl benzoic acid 2,3,4,5,6-pentafluorophenyl ester
PubChem CID 24229473
SMILES CN1C=CC(=N1)C1=CC=C(C=C1)C(=O)OC1=C(F)C(F)=C(F)C(F)=C1F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 2,3,4,5,6-pentafluorophenyl 4-(1-methyl-1H-pyrazol-3-yl)benzoate
Formula Weight 368.26
Percent Purity 97%
Chemical Name or Material Pentafluorophenyl 4-(1-methyl-1H-pyrazol-3-yl)benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.