missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentafluorophenyl 4-(1-methyl-1H-pyrazol-3-yl)benzoate, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
IUPAC Name | (2,3,4,5,6-pentafluorophenyl) 4-(1-methylpyrazol-3-yl)benzoate |
Formula Weight | 368.26 |
Percent Purity | 97% |
Chemical Name or Material | Pentafluorophenyl 4-(1-methyl-1H-pyrazol-3-yl)benzoate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'