missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl 2-morpholino-5-(trifluoromethyl)benzoate, 97%, Thermo Scientific™

Product Code. 10567464
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10567464 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10567464 Supplier Thermo Scientific Maybridge Supplier No. CC57826DA
 Restricted Item
Estimated Shipment: 22-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 898289-52-6
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C18H11F8NO3
MDL Number MFCD09025890
Quantity 1 g
Synonym pentafluorophenyl 2-morpholino-5-trifluoromethyl benzoate, pentafluorophenyl 2-morpholin-4-yl-5-trifluoromethyl benzoate, 2,3,4,5,6-pentafluorophenyl 2-morpholin-4-yl-5-trifluoromethyl benzoate, 4-morpholin-4-yl-3-pentafluorophenoxy carbonyl benzotrifluoride, 2,3,4,5,6-pentakis fluoranyl phenyl 2-morpholin-4-yl-5-trifluoromethyl benzoate, 2-4-morpholinyl-5-trifluoromethyl benzoic acid 2,3,4,5,6-pentafluorophenyl ester
InChI Key NRNRYELFJREPET-UHFFFAOYSA-N
SMILES C1COCCN1C2=C(C=C(C=C2)C(F)(F)F)C(=O)OC3=C(C(=C(C(=C3F)F)F)F)F
IUPAC Name (2,3,4,5,6-pentafluorophenyl) 2-morpholin-4-yl-5-(trifluoromethyl)benzoate
Molecular Weight (g/mol) 441.277
PubChem CID 24229574
Formula Weight 441.27
Percent Purity 97%
Chemical Name or Material Pentafluorophenyl 2-morpholino-5-(trifluoromethyl)benzoate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.