missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl 1-methyl-1H-indole-7-carboxylate, 97%, Thermo Scientific™

Product Code. 10576914
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 941716-96-7
Molecular Formula C16H8F5NO2
Molecular Weight (g/mol) 341.24
MDL Number MFCD09879962
InChI Key UEOKRSIJVAQHID-UHFFFAOYSA-N
Synonym pentafluorophenyl 1-methyl-1h-indole-7-carboxylate, pentafluorophenyl 1-methylindole-7-carboxylate, 2,3,4,5,6-pentakis fluoranyl phenyl 1-methylindole-7-carboxylate, 1-methyl-7-indolecarboxylic acid 2,3,4,5,6-pentafluorophenyl ester
PubChem CID 24229744
IUPAC Name (2,3,4,5,6-pentafluorophenyl) 1-methylindole-7-carboxylate
SMILES CN1C=CC2=CC=CC(C(=O)OC3=C(F)C(F)=C(F)C(F)=C3F)=C12

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 341.23
Percent Purity 97%
Chemical Name or Material Pentafluorophenyl 1-methyl-1H-indole-7-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'