missing translation for 'onlineSavingsMsg'
Learn More

(Pentafluoroethyl)trimethylsilane, 97%

Product Code. 15500624 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15500624 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15500624 Supplier Thermo Scientific Acros Supplier No. 459300250
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 124898-13-1 | C5H9F5Si | 192.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124898-13-1
Molecular Formula C5H9F5Si
Molecular Weight (g/mol) 192.2
InChI Key MTPVUVINMAGMJL-UHFFFAOYSA-N
Synonym pentafluoroethyl trimethylsilane, trimethyl pentafluoroethyl silane, pentafluoroethyltrimethylsilane, trimethylpentafluoroethylsilane, trimethyl perfluoroethyl silane, trimethyl 1,1,2,2,2-pentafluoroethyl silane, silane, trimethyl pentafluoroethyl, silane,trimethyl 1,1,2,2,2-pentafluoroethyl, acmc-20aple, trimethylsilylpentafluoroethane
PubChem CID 2760820
IUPAC Name trimethyl(1,1,2,2,2-pentafluoroethyl)silane
SMILES C[Si](C)(C)C(C(F)(F)F)(F)F

Specifications

Density 1.0500g/mL
Boiling Point 70.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Linear Formula F3CCF2Si(CH3)3
Quantity 25 g
Specific Gravity 1.05
Solubility Information Solubility in water: immiscible.
Formula Weight 192.2
Percent Purity 97%
Chemical Name or Material (Pentafluoroethyl)trimethylsilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (Pentafluoroethyl)trimethylsilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.