missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorobenzoyl chloride, 98%

Product Code. p-4510036 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10466780

Brand: Thermo Scientific Acros 147120050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2251-50-5 | C7ClF5O | 230.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Acylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2251-50-5
Molecular Formula C7ClF5O
Molecular Weight (g/mol) 230.52
MDL Number MFCD00000657
InChI Key MYHOHFDYWMPGJY-UHFFFAOYSA-N
Synonym pentafluorobenzoyl chloride, perfluorobenzoyl chloride, benzoyl chloride, pentafluoro, pentafluorobenzoylchloride, pubchem15522, acmc-1covi, intermediates-zcf02601, pentafluorobenzoic acid chloride, attercop-chm at133861, 67924-15-6 unspecified acetate
PubChem CID 75256
ChEBI CHEBI:39425
SMILES FC1=C(F)C(F)=C(C(Cl)=O)C(F)=C1F

Specifications

Color Colorless to Yellow
Density 1.6000g/mL
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6F5COCl
Refractive Index 1.4520 to 1.4540
Quantity 5 g
Specific Gravity 1.6
Solubility Information Solubility in water: decomposes.
IUPAC Name 2,3,4,5,6-pentafluorobenzoyl chloride
Formula Weight 230.52
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Pentafluorobenzoyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • Pentafluorobenzoyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.