missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorobenzoyl chloride, 98%

Product Code. 11449613
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449613 25 g 25g
11439613 5 g 5g
11459613 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11449613 Supplier Thermo Scientific Alfa Aesar Supplier No. A12420.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2251-50-5 | C7ClF5O | 230.52 g/mol

A reagent used for deriving primary amines. 2,3,4,5,6-Pentafluorobenzoyl chloride was used for derivatization of 5-androstenediol, testosterone, estradiol , dihydrotestosterone and estrone in a study to determine their serum levels. 2,3,4,5,6-Pentafluorobenzoyl chloride was used to derivative dimethylamine(DMA) in study to develop a rapid and accurate GC-MS method for determination of DMA in human urine. It was used in a study to detect the presence of Enterobacter cloacae in cultures of Leuconostoc mesenteroide by gas chromatography/mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A reagent used for deriving primary amines. 2,3,4,5,6-Pentafluorobenzoyl chloride was used for derivatization of 5-androstenediol, testosterone, estradiol , dihydrotestosterone and estrone in a study to determine their serum levels. 2,3,4,5,6-Pentafluorobenzoyl chloride was used to derivative dimethylamine(DMA) in study to develop a rapid and accurate GC-MS method for determination of DMA in human urine. It was used in a study to detect the presence of Enterobacter cloacae in cultures of Leuconostoc mesenteroide by gas chromatography/mass spectrometry.

Solubility
Decomposes in water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, Water, Alcohols, Strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2251-50-5
Molecular Formula C7ClF5O
Molecular Weight (g/mol) 230.52
MDL Number MFCD00000657
InChI Key MYHOHFDYWMPGJY-UHFFFAOYSA-N
Synonym pentafluorobenzoyl chloride, perfluorobenzoyl chloride, benzoyl chloride, pentafluoro, pentafluorobenzoylchloride, pubchem15522, acmc-1covi, intermediates-zcf02601, pentafluorobenzoic acid chloride, attercop-chm at133861, 67924-15-6 unspecified acetate
PubChem CID 75256
ChEBI CHEBI:39425
IUPAC Name 2,3,4,5,6-pentafluorobenzoyl chloride
SMILES FC1=C(F)C(F)=C(C(Cl)=O)C(F)=C1F

Specifications

Density 1.668
Boiling Point 158°C to 159°C
Refractive Index 1.4535
Quantity 25 g
UN Number UN3265
Beilstein 2054397
Sensitivity Moisture sensitive
Solubility Information Decomposes in water.
Formula Weight 230.52
Percent Purity 98%
Chemical Name or Material Pentafluorobenzoyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.