missing translation for 'onlineSavingsMsg'
Learn More

Pentafluoroaniline, 97%

Product Code. 11433957
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433957 100 g 100g
11413957 5 g 5g
11423957 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11433957 Supplier Thermo Scientific Alfa Aesar Supplier No. A19348.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 771-60-8 | C6H2F5N | 183.08 g/mol

2,3,4,5,6-Pentafluoroaniline finds it uses in the preparation of pentafluorophenylammonium triflate, an efficient catalyst for esterification and thioesterification. It is also used in the synthesis of various titanium complexes having two anionic [N, O-] bidentate salicylaldiminato ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,4,5,6-Pentafluoroaniline finds it uses in the preparation of pentafluorophenylammonium triflate, an efficient catalyst for esterification and thioesterification. It is also used in the synthesis of various titanium complexes having two anionic [N, O-] bidentate salicylaldiminato ligands.

Solubility
Soluble in toluene, and organic solvents. Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 771-60-8
Molecular Formula C6H2F5N
Molecular Weight (g/mol) 183.08
MDL Number MFCD00007643
InChI Key NOXLGCOSAFGMDV-UHFFFAOYSA-N
Synonym pentafluoroaniline, pentafluorophenylamine, aminopentafluorobenzene, benzenamine, 2,3,4,5,6-pentafluoro, 2,3,4,5,6-pentafluorobenzenamine, aniline, 2,3,4,5,6-pentafluoro, unii-ms06il3y2q, ms06il3y2q, 2,3,4,5,6-pentafluorophenylamine, pubchem1546
PubChem CID 13040
SMILES NC1=C(F)C(F)=C(F)C(F)=C1F

Specifications

Melting Point 34°C to 37°C
Density 1.744
Boiling Point 156°C to 157°C
Flash Point 73°C (163°F)
Quantity 100 g
Beilstein 1819387
Solubility Information Soluble in toluene,and organic solvents. Insoluble in water.
IUPAC Name pentafluoroaniline
Formula Weight 183.08
Percent Purity ≥98%
Chemical Name or Material Pentafluoroaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.