missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentaethylene Glycol Dimethanesulfonate, TRC
Specifications
Specifications
| Chemical Name or Material | Pentaethylene Glycol Dimethanesulfonate |
| CAS | 109789-39-1 |
| Molecular Formula | C10 H22 O6 . 2 C H4 O3 S |
| Quantity | 250 mg |
| Synonym | 3,6,9,12-Tetraoxatetradecane-1,14-diol 1,14-Dimethanesulfonate, Pentaethylene Glycol Dimesylate |
| InChI Formula | InChI=1S/C10H22O6.2CH4O3S/c11-1-3-13-5-7-15-9-10-16-8-6-14-4-2-12;2*1-5(2,3)4/h11-12H,1-10H2;2*1H3,(H,2,3,4) |
| SMILES | CS(=O)(=O)O.CS(=O)(=O)O.OCCOCCOCCOCCOCCO |
| IUPAC Name | 2-[2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethoxy]ethanol;methanesulfonic acid |
| Molecular Weight (g/mol) | 430.489 |
| Formula Weight | 430.118 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?