missing translation for 'onlineSavingsMsg'
Learn More

Pentaethylene Glycol Dimethanesulfonate, TRC

Product Code. 30426565
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30426565 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30426565 Supplier Toronto Research Chemicals Supplier No. TRCP2702501G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pentaethylene Glycol Dimethanesulfonate
CAS 109789-39-1
Molecular Formula C10 H22 O6 . 2 C H4 O3 S
Quantity 1 g
Synonym 3,6,9,12-Tetraoxatetradecane-1,14-diol 1,14-Dimethanesulfonate, Pentaethylene Glycol Dimesylate
InChI Formula InChI=1S/C10H22O6.2CH4O3S/c11-1-3-13-5-7-15-9-10-16-8-6-14-4-2-12;2*1-5(2,3)4/h11-12H,1-10H2;2*1H3,(H,2,3,4)
SMILES CS(=O)(=O)O.CS(=O)(=O)O.OCCOCCOCCOCCOCCO
IUPAC Name 2-[2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethoxy]ethanol;methanesulfonic acid
Molecular Weight (g/mol) 430.489
Formula Weight 430.118
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.