missing translation for 'onlineSavingsMsg'
Learn More

Pentaethylene glycol, 98+%

Product Code. 11429392
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429392 10 g 10g
11439392 50 g 50g
11449392 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11429392 Supplier Thermo Scientific Alfa Aesar Supplier No. A10133.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as an intermediate in the synthesis of cyclic and noncyclic polyethers, complexing agents and immobilized phase-transfer catalysts. It acts as a catalyst for potassium hydroxide induced elimination reactions in benzene. Further, it is used in functional fluids and viscosity adjustors. | CAS: 4792-15-8 | C10H22O6 | 238.28 g/mol

Pentaethylene glycol is used as an intermediate in the synthesis of cyclic and noncyclic polyethers, complexing agents and immobilized phase-transfer catalysts. It acts as a catalyst for potassium hydroxide induced elimination reactions in benzene. Further, it is used in functional fluids and viscosity adjustors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentaethylene glycol is used as an intermediate in the synthesis of cyclic and noncyclic polyethers, complexing agents and immobilized phase-transfer catalysts. It acts as a catalyst for potassium hydroxide induced elimination reactions in benzene. Further, it is used in functional fluids and viscosity adjustors.

Solubility
Miscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, acid chlorides and acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4792-15-8
Molecular Formula C10H22O6
Molecular Weight (g/mol) 238.28
MDL Number MFCD00002878
InChI Key JLFNLZLINWHATN-UHFFFAOYSA-N
Synonym pentaethylene glycol, 3,6,9,12-tetraoxatetradecane-1,14-diol, pentaethyleneglycol, pentaglycol, ho ch2ch2o 5h, 3,6,9,12-tetraoxatetradocane-1,14-diol, 2-2-2-2-2-hydroxyethoxy ethoxy ethoxy ethoxy ethanol, dsstox_cid_7579, dsstox_rid_78510, dsstox_gsid_27579
PubChem CID 62551
ChEBI CHEBI:39631
IUPAC Name 2-[2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethoxy]ethanol
SMILES C(COCCOCCOCCOCCO)O

Specifications

Density 1.126 g/mL
Boiling Point 338°C to 340°C
Flash Point >110°C (230°F)
Assay Percent Range >98%
Refractive Index 1.462
Quantity 10 g
Beilstein 1635593
Solubility Information Miscible with water.
Formula Weight 238.28
Chemical Name or Material Pentaethylene glycol
compliance-icons
Produktbeteckning
  • Pentaethylene glycol
Signalord
  • Warning
Riskkategorier
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faroangivelser
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Skyddsangivelse
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Kompletterande information
  • MIXTURE LIST-Contain: 3,6,9,12-Tetraoxatetradecane-1,14-diol

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.